MAT333121630 Matrix Scientific

1-(2-Nitrophenyl)-1H-pyrrole-2,5-dione

  • MDL number MFCD00022565
  • Cas Number : 2973-15-1
  • Molecular Weight : 218.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/066471.html

Safety Information

MSDS http://www.matrixscientific.com/066471.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart