MAT333121676 Matrix Scientific

3-(3,4-Dimethoxyphenyl)-7-hydroxy-4H-chromen-4-one

  • MDL number MFCD00076013
  • Cas Number : 24160-14-3
  • Molecular Weight : 298.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/066517.html

Safety Information

MSDS http://www.matrixscientific.com/066517.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart