MAT333121694 Matrix Scientific

(2Z)-2-Benzylidene-6-hydroxy-1-benzofuran-3(2H)-one

  • MDL number MFCD04145101
  • Cas Number : 139276-16-7
  • Molecular Weight : 238.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 262
Country of Origin USA
Product Link http://www.matrixscientific.com/066535.html

Safety Information

MSDS http://www.matrixscientific.com/066535.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart