MAT338367396 Matrix Scientific

4-Amino-2-bromobenzonitrile

  • MDL number MFCD03695814
  • Cas Number : 53312-82-6
  • Molecular Weight : 197.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.matrixscientific.com/066623.html

Safety Information

hcodes Toxic
MSDS http://www.matrixscientific.com/066623.html
Availability details for
Quantity
  • Availability
  • Add to Cart