MAT338367405 Matrix Scientific

2-Methyl-5-(trifluoromethyl)benzaldehyde

  • MDL number MFCD06660200
  • Cas Number : 886498-85-7
  • Molecular Weight : 188.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/066632.html

Safety Information

MSDS http://www.matrixscientific.com/066632.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart