MAT338367425 Matrix Scientific

1-Ethyl-3-methyl-4-nitro-1H-pyrazole-5-carboxylic acid

  • MDL number MFCD00207991
  • Cas Number : 26308-39-4
  • Molecular Weight : 199.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/066654.html

Safety Information

MSDS http://www.matrixscientific.com/066654.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart