MAT338367450 Matrix Scientific

2-Cyclopropylbenzonitrile

  • MDL number MFCD00463731
  • Cas Number : 3154-99-2
  • Molecular Weight : 143.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/066679.html

Safety Information

MSDS http://www.matrixscientific.com/066679.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart