MAT338367479 Matrix Scientific

5-Piperazin-1-yl-1,6-naphthyridine hydrochloride

  • MDL number MFCD09753939
  • Cas Number : 1171412-23-9
  • Molecular Weight : 250.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin USA
Product Link http://www.matrixscientific.com/066708.html

Safety Information

MSDS http://www.matrixscientific.com/066708.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart