MAT338367496 Matrix Scientific

sec-Butylhydrazine dihydrochloride

  • MDL number MFCD11840328
  • Cas Number : 1177361-36-2
  • Molecular Weight : 161.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/066725.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/066725.html
Availability details for
Quantity
  • Availability
  • Add to Cart