MAT338367612 Matrix Scientific

3-Ethylpiperidine hydrochloride

  • MDL number MFCD19442002
  • Cas Number : 13603-10-6
  • Molecular Weight : 149.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 142
Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/053683.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/053683.html
Availability details for
Quantity
  • Availability
  • Add to Cart