MAT338367633 Matrix Scientific

3-(5-Chloropyridin-2-yl)-3-oxopropanenitrile

  • MDL number MFCD15526709
  • Cas Number : 1107053-87-1
  • Molecular Weight : 180.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 121-123
Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/066876.html

Safety Information

MSDS http://www.matrixscientific.com/066876.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart