MAT338367683 Matrix Scientific

Methyl 4-(pyrrolidin-1-ylmethyl)benzoate

  • MDL number MFCD01452504
  • Cas Number : 160598-45-8
  • Molecular Weight : 219.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/066929.html

Safety Information

MSDS http://www.matrixscientific.com/066929.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart