MAT343137439 Matrix Scientific

2-Oxa-6-azaspiro[3.4]octane

  • MDL number MFCD14586455
  • Cas Number : 220290-68-6
  • Molecular Weight : 113.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Density 1.08
Flash Point 63
Country of Origin USA
Boiling point 187
Product Link http://www.matrixscientific.com/067048.html

Safety Information

hcodes Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart