MAT359313401 Matrix Scientific

Ethyl 2-amino-4-chlorothieno[2,3-d]-pyrimidine-6-carboxylate

  • MDL number MFCD18074287
  • Cas Number : 847560-46-7
  • Molecular Weight : 257.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 253-255
Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/067162.html

Safety Information

MSDS http://www.matrixscientific.com/067162.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart