MAT359313407 Matrix Scientific

Ethyl 4-chloro-5-nitro-6-oxo-1,6-dihydropyridine-3-carboxylate

  • MDL number MFCD20921575
  • Cas Number : 1210835-74-7
  • Molecular Weight : 246.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 176-178
Country of Origin USA
Product Link http://www.matrixscientific.com/067168.html

Safety Information

MSDS http://www.matrixscientific.com/067168.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart