MAT359313408 Matrix Scientific

Ethyl 4,5-dichloro-1-methyl-6-oxo-1,6-dihydropyridine-3-carboxylate

  • MDL number MFCD09031524
  • Cas Number : 853105-72-3
  • Molecular Weight : 250.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 132-134
Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/067169.html

Safety Information

MSDS http://www.matrixscientific.com/067169.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart