MAT364046984 Matrix Scientific

3-Phenyl-decahydro-1,4-diaza-cyclopenta-cyclooctene-4-carboxylic acid tert-butyl ester

  • MDL number MFCD21605790
  • Cas Number : 2098497-07-3
  • Molecular Weight : 330.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/067412.html

Safety Information

MSDS http://www.matrixscientific.com/067412.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart