MAT364047096 Matrix Scientific

Azetidine-3-carboxylic acid [2-(1-methyl-1H-benzoimidazol-2-yl)-ethyl]-amide hydrochloride

  • MDL number MFCD21605908
  • Cas Number : 1361112-47-1
  • Molecular Weight : 294.79

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/067624.html

Safety Information

MSDS http://www.matrixscientific.com/067624.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart