MAT364047204 Matrix Scientific

4-Acetyl-piperazine-2-carboxylic acid methylamide hydrochloride

  • MDL number MFCD21606025
  • Cas Number : 1361111-27-4
  • Molecular Weight : 221.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/067913.html

Safety Information

MSDS http://www.matrixscientific.com/067913.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart