MAT364047221 Matrix Scientific

1-Methyl-piperazine-2-carboxylic acid methylamide dihydrochloride

  • MDL number MFCD21606043
  • Cas Number : 1361111-35-4
  • Molecular Weight : 230.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/068004.html

Safety Information

MSDS http://www.matrixscientific.com/068004.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart