MAT364047231 Matrix Scientific

1-Methyl-piperazine-2-carboxylic acid amide dihydrochloride

  • MDL number MFCD21606053
  • Cas Number : 859213-28-8
  • Molecular Weight : 216.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/068029.html

Safety Information

MSDS http://www.matrixscientific.com/068029.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart