MAT364047328 Matrix Scientific

3-Pyrrolidin-2-yl-1H-pyrazole dihydrochloride

  • MDL number MFCD21606153
  • Cas Number : 1361114-72-8
  • Molecular Weight : 210.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/068204.html

Safety Information

MSDS http://www.matrixscientific.com/068204.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart