MAT364047359 Matrix Scientific

2-Methoxy-N-(2-piperidin-3-yl-ethyl)-acetamide hydrochloride

  • MDL number MFCD21606185
  • Cas Number : 1361116-55-3
  • Molecular Weight : 236.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/068244.html

Safety Information

MSDS http://www.matrixscientific.com/068244.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart