MAT364047535 Matrix Scientific

2-(1-Methanesulfonyl-piperidin-4-yl)-ethylamine hydrochloride

  • MDL number MFCD09455630
  • Cas Number : 1158209-14-3
  • Molecular Weight : 242.77

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/067823.html

Safety Information

MSDS http://www.matrixscientific.com/067823.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart