MAT364047553 Matrix Scientific

3-Methyl-3,4,7,8-tetrahydro-2,5(1H,6H)-quinolinedione

  • MDL number MFCD13184135
  • Cas Number : 77903-18-5
  • Molecular Weight : 179.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 206-208
Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/043187.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/043187.html
Availability details for
Quantity
  • Availability
  • Add to Cart