MAT364047561 Matrix Scientific

N-Benzyl-N-ethyl-cyclohexane-1,4-diamine hydrochloride

  • MDL number MFCD16621912
  • Cas Number : 1229623-48-6
  • Molecular Weight : 268.83

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/067849.html

Safety Information

MSDS http://www.matrixscientific.com/067849.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart