MAT364047723 Matrix Scientific

4-Phenethyl-piperazine-2-carboxylic acid amide dihydrochloride

  • MDL number MFCD16621931
  • Cas Number : 1313254-64-6
  • Molecular Weight : 306.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/068011.html

Safety Information

MSDS http://www.matrixscientific.com/068011.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart