MAT364047853 Matrix Scientific

4-(2-Fluoro-phenyl)-3-oxo-butyric acid methyl ester

  • MDL number MFCD08457131
  • Cas Number : 1048917-90-3
  • Molecular Weight : 210.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/067351.html

Safety Information

MSDS http://www.matrixscientific.com/067351.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart