MAT364048114 Matrix Scientific

Dimethyl-(4-pyrrolidin-3-yl-pyrimidin-2-yl)-amine dihydrochloride

  • MDL number MFCD21606244
  • Cas Number : 1361113-37-2
  • Molecular Weight : 265.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/068317.html

Safety Information

MSDS http://www.matrixscientific.com/068317.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart