MAT364048158 Matrix Scientific

1-(5-Bromo-3',4',5',6'-tetrahydro-2'H-[2,4']bipyridinyl-1'-yl)-ethanone

  • MDL number MFCD19691509
  • Cas Number : 1316221-39-2
  • Molecular Weight : 283.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/068347.html

Safety Information

MSDS http://www.matrixscientific.com/068347.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart