MAT364048169 Matrix Scientific

4-(3-Methyl-3H-imidazol-4-yl)-piperidine dihydrochloride

  • MDL number MFCD11111436
  • Cas Number : 147960-50-7
  • Molecular Weight : 238.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/068278.html

Safety Information

MSDS http://www.matrixscientific.com/068278.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart