MAT364048923 Matrix Scientific

tert-Butyl 4-(5-chloro-1H-indazol-3-yl)piperidine-1-carboxylate

  • MDL number MFCD12912623
  • Cas Number : 1198284-74-0
  • Molecular Weight : 335.84

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/069115.html

Safety Information

MSDS http://www.matrixscientific.com/069115.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart