MAT364049172 Matrix Scientific

N,N,3-Trimethylazetidin-3-amine

  • MDL number MFCD17170039
  • Cas Number : 149696-13-9
  • Molecular Weight : 114.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/069362.html

Safety Information

MSDS http://www.matrixscientific.com/069362.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart