MAT364050090 Matrix Scientific

1-(2-Chloroacetyl)-5,11-dihydro-pyrido-[2,3-b][1,4]benzodiazepin-6-one

  • MDL number MFCD21648226
  • Cas Number : 28797-48-0
  • Molecular Weight : 289.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 212
Purity 98
Density 1.415
Country of Origin USA
Boiling point 481
Product Link http://www.matrixscientific.com/070857.html

Safety Information

MSDS http://www.matrixscientific.com/070857.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart