MAT364050454 Matrix Scientific

tert-Butyl 2-oxo-1,7-diazaspiro-[4.4]nonane-7-carboxylate

  • MDL number MFCD12198506
  • Cas Number : 1160246-72-9
  • Molecular Weight : 240.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/070619.html

Safety Information

MSDS http://www.matrixscientific.com/070619.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart