MAT364050514 Matrix Scientific

2-Thia-6-azaspiro[3.3]heptane-2,2-dioxide

  • MDL number MFCD17017544
  • Cas Number : 1263182-09-7
  • Molecular Weight : 147.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/070679.html

Safety Information

MSDS http://www.matrixscientific.com/070679.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart