MAT364050577 Matrix Scientific

Ethyl 2-(1,3-benzothiazol-2-yl)-2-oxoacetate

  • MDL number MFCD19443897
  • Cas Number : 20474-55-9
  • Molecular Weight : 235.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/070742.html

Safety Information

MSDS http://www.matrixscientific.com/070742.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart