MAT364050611 Matrix Scientific

(E)-(3-Chloroprop-1-enyl)-4-bromobenzene

  • MDL number MFCD01861828
  • Cas Number : 104293-07-4
  • Molecular Weight : 231.53

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 60-62
Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/070778.html

Safety Information

MSDS http://www.matrixscientific.com/070778.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart