MAT364050664 Matrix Scientific

4'-Fluoro-2'-methylacetophenone

  • MDL number MFCD00028041
  • Cas Number : 446-29-7
  • Molecular Weight : 152.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.matrixscientific.com/070852.html

Safety Information

MSDS http://www.matrixscientific.com/070852.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart