MAT369262690 Matrix Scientific

2,3-Difluoro-4-(difluoromethyl)pyridine

  • MDL number MFCD22199287
  • Cas Number : 1374659-38-7
  • Molecular Weight : 165.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/071389.html

Safety Information

MSDS http://www.matrixscientific.com/071389.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart