MAT369262691 Matrix Scientific

5-Bromo-2-chloro-4-(difluoromethyl)pyridine

  • MDL number MFCD22199288
  • Cas Number : 1374659-34-3
  • Molecular Weight : 242.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/071390.html

Safety Information

MSDS http://www.matrixscientific.com/071390.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart