MAT369262713 Matrix Scientific

2-Chloro-3-(difluoromethyl)-5-fluoropyridine

  • MDL number MFCD22199309
  • Cas Number : 1374659-31-0
  • Molecular Weight : 181.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/071603.html

Safety Information

MSDS http://www.matrixscientific.com/071603.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart