MAT369262752 Matrix Scientific

Ethyl 4-amino-1H-pyrazole-5-carboxylate hydrochloride

  • MDL number MFCD09759000
  • Cas Number : 55904-61-5
  • Molecular Weight : 191.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/031130.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/031130.html
Availability details for
Quantity
  • Availability
  • Add to Cart