MAT369262901 Matrix Scientific

1,2-Bis-(trifluoromethyl)benzene

  • MDL number MFCD03094218
  • Cas Number : 433-95-4
  • Molecular Weight : 214.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Boiling point 140-142
Product Link http://www.matrixscientific.com/071471.html

Safety Information

MSDS http://www.matrixscientific.com/071471.html
pictograms Flammable
Availability details for
Quantity
  • Availability
  • Add to Cart