MAT369262912 Matrix Scientific

(2-Chloro-1,1-difluoroethyl)benzene

  • MDL number MFCD14525555
  • Cas Number : 55805-08-8
  • Molecular Weight : 176.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/071482.html

Safety Information

MSDS http://www.matrixscientific.com/071482.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart