MAT369262923 Matrix Scientific

3,5-Dimethylbenzotrifluoride

  • MDL number MFCD12026302
  • Cas Number : 86845-29-6
  • Molecular Weight : 174.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/071493.html

Safety Information

MSDS http://www.matrixscientific.com/071493.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart