MAT369262950 Matrix Scientific

4,5-Bis(trifluoromethyl)imidazole

  • MDL number MFCD12546953
  • Cas Number : 651-34-3
  • Molecular Weight : 204.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 165-168
Purity 97
Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/071520.html

Safety Information

MSDS http://www.matrixscientific.com/071520.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart