MAT369262989 Matrix Scientific

2-Methyl-4-(trifluoromethyl)pyridine

  • MDL number MFCD13185742
  • Cas Number : 106877-17-2
  • Molecular Weight : 161.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/071560.html

Safety Information

MSDS http://www.matrixscientific.com/071560.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart