MAT369262994 Matrix Scientific

1,3-Dichloro-2,4-bis(trifluoromethyl)benzene

  • MDL number MFCD16140187
  • Cas Number : 1221272-88-3
  • Molecular Weight : 283

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Boiling point 98� torr
Product Link http://www.matrixscientific.com/071565.html

Safety Information

MSDS http://www.matrixscientific.com/071565.html
pictograms Toxic
Availability details for
Quantity
  • Availability
  • Add to Cart