MAT370169758 Matrix Scientific

2-Oxa-5-azaspiro[3.4]octane hemioxalate

  • MDL number MFCD22373635
  • Cas Number : 1380571-82-3
  • Molecular Weight : 316.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/067047.html

Safety Information

MSDS http://www.matrixscientific.com/067047.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart